Anti-ASNA1 Rabbit Polyclonal Antibody (Cy5.5®)
Lieferant: Bioss
BS-7972R-CY5.5
BOSSBS-7972R-CY5.5EA 900 EUR
BOSSBS-7972R-CY5.5
Anti-ASNA1 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
ASNA1 is the human homolog of the E.coli arsA gene which is an ATPase, and is the catalytic component of a multisubunit oxyanion pump responsible for resistance to arsenicals and antimonials.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ASNA1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat