Anti-CDKAL1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-7997R-A488
BOSSBS-7997R-A488EA 893 EUR
BOSSBS-7997R-A488
Anti-CDKAL1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Catalyzes the methylthiolation of N6-threonylcarbamoyladenosine (t(6)A), leading to the formation of 2-methylthio-N6-threonylcarbamoyladenosine (ms(2)t(6)A) at position 37 in tRNAs that read codons beginning with adenine.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CDKAL1
Clonality: Polyclonal
Clone: 0
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR