Anti-CDKAL1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-7997R-A680
BOSSBS-7997R-A680EA 893 EUR
BOSSBS-7997R-A680
Anti-CDKAL1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Catalyzes the methylthiolation of N6-threonylcarbamoyladenosine (t(6)A), leading to the formation of 2-methylthio-N6-threonylcarbamoyladenosine (ms(2)t(6)A) at position 37 in tRNAs that read codons beginning with adenine.
Type: Primary
Antigen: CDKAL1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 375-460/579
Host: Rabbit
Isotype: IgG
Reactivity: Rat