Anti-ICMT Rabbit Polyclonal Antibody (Cy3®)
Lieferant: Bioss
BS-8019R-CY3
BOSSBS-8019R-CY3EA 900 EUR
BOSSBS-8019R-CY3
Anti-ICMT Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Catalyzes the post-translational methylation of isoprenylated C-terminal cysteine residues.Tissue specificity:Ubiquitously expressed. Expressed at higher levels in the cerebellum and putamen than in other brain regions. Abundant expression seen in the Purkinje cells and pontine neurons.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ICMT
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat