Anti-ICMT Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-8019R-CY5.5
BOSSBS-8019R-CY5.5EA 900 EUR
BOSSBS-8019R-CY5.5
Anti-ICMT Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Catalyzes the post-translational methylation of isoprenylated C-terminal cysteine residues.Tissue specificity:Ubiquitously expressed. Expressed at higher levels in the cerebellum and putamen than in other brain regions. Abundant expression seen in the Purkinje cells and pontine neurons.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ICMT
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR