Anti-CNKSR1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-8028R-A555
BOSSBS-8028R-A555EA 893 EUR
BOSSBS-8028R-A555
Anti-CNKSR1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
This gene is a necessary element in receptor tyrosine kinase pathways, possibly as a tyrosine phosphorylation target. It is involved in regulation of RAF in the MAPK pathway and may also play a role in a MAPK-independent pathway. [provided by RefSeq, Jul 2008].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CNKSR1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR