Anti-COMMD7 Rabbit Polyclonal Antibody
Lieferant: Bioss
BS-8036R
BOSSBS-8036REA 725 EUR
BOSSBS-8036R
Anti-COMMD7 Rabbit Polyclonal Antibody
Antikörper
COMMD is a new family of proteins with homology to MURR1, a multifunctional protein that inhibits NFkB. These proteins form multimeric complexes and were identified in a biochemical screen for MURR1-associated factors. The family is defined by the presence of a conserved and unique motif termed the COMM (copper metabolism gene MURR1) domain, which functions as an interface for protein-protein interactions. The prototype of this family, MURR1/COMMD1, suppresses NFkB by affecting the association of NF-kappaB with chromatin. COMMD7 (COMM domain-containing protein 7) interacts with COMMD1 via its COMM domain. It also associates with the NF-kappa-B complex and suppresses its transcriptional activity.
IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: COMMD7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen