Anti-KLHL3 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-8053R-CY5.5
BOSSBS-8053R-CY5.5EA 900 EUR
BOSSBS-8053R-CY5.5
Anti-KLHL3 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
KLHL3 protein contains a poxvirus and zinc finger domain at the N-terminus and six tandem repeats (kelch repeats) at the C-terminus. At the amino acid level, KLHL3 shares 77% similarity with Drosophila kelch and 89% similarity with Mayven (KLHL2), another human kelch homolog. At least three isoforms are produced and may be the result of alternative promoter usage. The KLHL3 maps within the smallest commonly deleted segment in myeloid leukemias characterized by a deletion of 5q; however, no inactivating mutations of KLHL3 could be detected in malignant myeloid disorders with loss of 5q.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KLHL3
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR