Anti-CCDC65 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-8100R-CY3
BOSSBS-8100R-CY3EA 893 EUR
BOSSBS-8100R-CY3
Anti-CCDC65 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
CCDC65 is a sperm tail protein that is posttranslationally modified during sperm capacitation (Zheng et al., 2006 [PubMed 17089017]).[supplied by OMIM, Mar 2008].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCDC65
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR