Anti-AVSD2 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-8184R
BOSSBS-8184REA 725 EUR
BOSSBS-8184R
Anti-AVSD2 Rabbit Polyclonal Antibody
Antikörper
CRELD1 is a protein composed of Epidermal growth factor-like repeats, which represent a group of cysteine-rich domains that mediate interactions between proteins of diverse function. Such domains are found in proteins that are either completely secreted or have transmembrane regions that link the protein to the cell surface.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: AVSD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR