Anti-CRYZL1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-8186R-A555
BOSSBS-8186R-A555EA 893 EUR
BOSSBS-8186R-A555
Anti-CRYZL1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
This gene encodes a protein that has sequence similarity to zeta crystallin, also known as quinone oxidoreductase. This zeta crystallin-like protein also contains an NAD(P)H binding site. Alternatively spliced transcript variants have been observed but their full-length nature has not been completely determined.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CRYZL1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR