Anti-CRYZL1 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-8186R-CY7
BOSSBS-8186R-CY7EA 900 EUR
BOSSBS-8186R-CY7
Anti-CRYZL1 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
This gene encodes a protein that has sequence similarity to zeta crystallin, also known as quinone oxidoreductase. This zeta crystallin-like protein also contains an NAD(P)H binding site. Alternatively spliced transcript variants have been observed but their full-length nature has not been completely determined.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CRYZL1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR