Anti-FAHD2A Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-8228R-CY3
BOSSBS-8228R-CY3EA 900 EUR
BOSSBS-8228R-CY3
Anti-FAHD2A Rabbit Polyclonal Antibody (Cy3®)
Antikörper
The FAH family contains two highly homologous 314 amino acid proteins, designated FAHD2A (fumarylacetoacetate hydrolase domain-containing protein 2A) and FAHD2B (fumarylacetoacetate hydrolase domain-containing protein 2A). FAHD2A and B utilize calcium and magnesium as cofactors, and may possess hydrolase activity. The genes encoding FAHD2A/B map to human chromosome 2, the second largest human chromosome, which consists of 237 million bases, encodes over 1,400 genes and makes up approximately 8% of the human genome. A number of genetic diseases are linked to genes on chromosome 2. Harlequin icthyosis, a rare and morbid skin deformity, is associated with mutations in the ABCA12 gene. The lipid metabolic disorder sitosterolemia is associated with ABCG5 and ABCG8. An extremely rare recessive genetic disorder, Alstré°‰ syndrome, is also associated with mutations to chromosome 2.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FAHD2A
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR