Anti-CTDSPL Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-8241R-CY5.5
BOSSBS-8241R-CY5.5EA 900 EUR
BOSSBS-8241R-CY5.5
Anti-CTDSPL Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Preferentially catalyzes the dephosphorylation of 'Ser-5' within the tandem 7 residues repeats in the C-terminal domain (CTD) of the largest RNA polymerase II subunit POLR2A. Negatively regulates RNA polymerase II transcription, possibly by controlling the transition from initiation/capping to processive transcript elongation (By similarity). Recruited by REST to neuronal genes that contain RE-1 elements, leading to neuronal gene silencing in non-neuronal cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CTDSPL
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR