Anti-DCAKD Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-8249R-A647
BOSSBS-8249R-A647EA 893 EUR
BOSSBS-8249R-A647
Anti-DCAKD Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
DCAKD belongs to the coaE family. It contains one DPCK (dephospho CoA kinase) domain. There are two isoforms.Coenzyme A (CoA) is an essential cofactor used in numerous biochemical pathways. It plays a critical role in the synthesis and oxidation of fatty acids and is vital to the citric acid cycle. The biosynthesis pathway of CoA from pantothenic acid (also known as vitamin B5) is essential and universal in prokaryotes and eukaryotes. In humans, the final steps of the biosynthesis pathway are carried out by the bifunctional enzyme COASY. The sequence of these enzymes are highly conserved between different bacterial species. The phosphopantetheine adenylyltransferase and decoenzyme A kinase activities of COASY are evolutionarily conserved activities. DCAKD (deCoA kinase domain containing protein) is a 231 amino acid protein that consists of a deCoA kinase domain and an ATP nucleotide binding motif. Localizing to mitochondria and the cytosol, DCAKD belongs to the coaE family which suggests that it may play a role in the biosynthesis of CoA.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DCAKD
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR