Anti-DCAKD Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-8249R-A680
BOSSBS-8249R-A680EA 893 EUR
BOSSBS-8249R-A680
Anti-DCAKD Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
DCAKD belongs to the coaE family. It contains one DPCK (dephospho CoA kinase) domain. There are two isoforms.Coenzyme A (CoA) is an essential cofactor used in numerous biochemical pathways. It plays a critical role in the synthesis and oxidation of fatty acids and is vital to the citric acid cycle. The biosynthesis pathway of CoA from pantothenic acid (also known as vitamin B5) is essential and universal in prokaryotes and eukaryotes. In humans, the final steps of the biosynthesis pathway are carried out by the bifunctional enzyme COASY. The sequence of these enzymes are highly conservedMore Product Information