Anti-DPYS Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-8287R-CY5.5
BOSSBS-8287R-CY5.5EA 900 EUR
BOSSBS-8287R-CY5.5
Anti-DPYS Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Catalyzes the second step of the reductive pyrimidine degradation, the reversible hydrolytic ring opening of dihydropyrimidines. Can catalyzes the ring opening of 5,6-dihydrouracil to N-carbamyl-alanine and of 5,6-dihydrothymine to N-carbamyl-amino isobutyrate.Tissue specificity:Liver and kidney.Involvement in disease:Defects in DPYS are the cause of dihydropyrimidinase deficiency (DHPD). DHPD is an autosomal recessive disorder characterized by dihydropyrimidinuria and associated with a variable clinical phenotype: epileptic or convulsive attacks, dysmorphic features and severe developmental delay, and congenital microvillous atrophy.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DPYS
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR