Anti-DPYD Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss
BS-8288R-FITC
BOSSBS-8288R-FITCEA 900 EUR
BOSSBS-8288R-FITC
Anti-DPYD Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Dihydropyrimidine dehydrogenase (DPYD) catalyzes the first rate-limiting step of the NADPH-dependent catabolism of uracil and thymine to dihydrouracil and dihydrothymine; thus, a deficiency of DPYD leads to an accumulation of uracil and thymine. Abnormal concentrations of these metabolites in bodily fluids may be the cause of neurological disease and a contraindication for treatment of cancer patients with certain pyrimidine analogs. DPYD also catalyzes the anticancer agent 5-fluorouracil (5-FU) pathway and is involved in the efficacy and toxicity of 5-FU. Variations in DPYD concentration may arise from alterations at the transcriptional level of the dihydropyrimidine dehydrogenase gene. Specifically, hypermethylation of the DPYD promoter downregulates dihydropyrimidine dehydrogenase expression. Deficient DPYD alleles may constitute a risk factor for severe toxicity following treatment with 5-FU.Involvement in disease:Defects in DPYD are the cause of dihydropyrimidine dehydrogenase deficiency (DPYD deficiency) ; also known as hereditary thymine-uraciluria or familial pyrimidinemia. DPYD deficiency is a disease characterized by persistent urinary excretion of excessive amounts of uracil, thymine and 5-hydroxymethyluracil. Patients suffering from this disease show a severe reaction to the anticancer drug 5-fluorouracil. This reaction includes stomatitis, Leukopenia, thrombocytopenia, hair loss, diarrhea, fever, marked weight loss, cerebellar ataxia, and neurologic symptoms, progressing to semicoma.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DPYD
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR