Anti-NDUFA9 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-8301R-CY3
BOSSBS-8301R-CY3EA 900 EUR
BOSSBS-8301R-CY3
Anti-NDUFA9 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NDUFA9
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR