Anti-HDHD1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Lieferant: Bioss
BS-8316R-A647
BOSSBS-8316R-A647EA 893 EUR
BOSSBS-8316R-A647
Anti-HDHD1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Dephosphorylates pseudouridine 5'-phosphate, a potential intermediate in rRNA degradation. Pseudouridine is then excreted intact in urine.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HDHD1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat