Anti-HDHD1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-8316R-A750
BOSSBS-8316R-A750EA 893 EUR
BOSSBS-8316R-A750
Anti-HDHD1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Dephosphorylates pseudouridine 5'-phosphate, a potential intermediate in rRNA degradation. Pseudouridine is then excreted intact in urine.

Type: Primary
Antigen: HDHD1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-100/228
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR