Anti-HDHD1 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-8316R-CY5.5
BOSSBS-8316R-CY5.5EA 900 EUR
BOSSBS-8316R-CY5.5
Anti-HDHD1 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Dephosphorylates pseudouridine 5'-phosphate, a potential intermediate in rRNA degradation. Pseudouridine is then excreted intact in urine.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HDHD1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR