Anti-HDHD2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-8317R-A680
BOSSBS-8317R-A680EA 893 EUR
BOSSBS-8317R-A680
Anti-HDHD2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
HDHD2 (haloacid dehalogenase-like hydrolase domain containing 2) is also known as DKFZp564D1378 and is a 259 amino acid protein that is expressed as two isoforms produced by alternative splicing. HDHD2 belongs to the HAD-like hydrolase superfamily, which contains a group of hydrolase enzymes that differ from the å/
Type: Primary
Antigen: HDHD2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1-100/259
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen