Anti-SERPINB13 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-8324R
BOSSBS-8324REA 719 EUR
BOSSBS-8324R
Anti-SERPINB13 Rabbit Polyclonal Antibody
Antikörper
Headpin is a skin-specific, UV-repressible serine proteinase inhibitor (serpin) belonging to the ovalbumin serpin family. Headpin is abundant in the human keratinocyte cell line HaCaT, and in lesional keratinocytes from psoriatic skin. Headpin downregulation occurs in squamous cell carcinoma of the oral cavity and in squamous cell carcinoma cell lines of the head and neck.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: SERPINB13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR