Anti-CPXCR1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-8331R-A350
BOSSBS-8331R-A350EA 893 EUR
BOSSBS-8331R-A350
Anti-CPXCR1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
CPXCR1 is widely expressed in fetal tissues, including the tongue, mandible and palate with an unknown function. Cleft palate most commonly occurs as a sporadic multifactorial disorder with a clear but difficult to define genetic component. As a semi-dominant disorder, X-linked cleft palate (CPX) provides a useful model to investigate a congenital defect that is little influenced by non-genetic factors.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CPXCR1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR