Anti-CPXCR1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-8331R-A750
BOSSBS-8331R-A750EA 893 EUR
BOSSBS-8331R-A750
Anti-CPXCR1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
CPXCR1 is widely expressed in fetal tissues, including the tongue, mandible and palate with an unknown function. Cleft palate most commonly occurs as a sporadic multifactorial disorder with a clear but difficult to define genetic component. As a semi-dominant disorder, X-linked cleft palate (CPX) provides a useful model to investigate a congenital defect that is little influenced by non-genetic factors.
Type: Primary
Antigen: CPXCR1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 151-250/301
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen