Anti-FGFR1OP Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-8345R-CY7
BOSSBS-8345R-CY7EA 900 EUR
BOSSBS-8345R-CY7
Anti-FGFR1OP Rabbit Polyclonal Antibody (Cy7®)
Antikörper
FOP, is a 399 amino acid protein that localizes to the centrosome and contains one LisH domain. Expressed ubiquitously with highest expression in kidney, heart, muscle, colon, liver, testis and pancreas, FOP functions as a homodimer that interacts with EB1 and CEP350 and is essential for anchoring microtubules to centrosomes. Chromosomal aberrations that involve the FOP gene are associated with the pathogenesis of stem cell myeloproliferative disorder (MPD), a condition that is characterized by eosinophilia and myeloid hyperplasia and ultimately leads to acute myeloid leukemia. FOP is expressed as multiple isoforms that are produced by alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FGFR1OP
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR