Anti-BPIL3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-8425R-A750
BOSSBS-8425R-A750EA 893 EUR
BOSSBS-8425R-A750
Anti-BPIL3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Members of the bactericidal/permeability-increasing protein family have antimicrobial properties and bind lipophilic substances, therefore targeting gram-negative bacteria. The bactericidal permeability increasing protein (BPI) is an antibacterial and endotoxin-neutralising molecule that is abundant in the granules of polymorphonuclear leukocytes (neutrophil granules). Sharing structural and sequence homologies with BPI, BPIL3 (Bactericidal/permeability-increasing protein-like 3) is a 453 amino acid secreted protein that contains the family?s common conserved feature of two cysteine residues that are critical for protein function. While BPIL3 is primarily expressed at low levels in tonsils, it has been found to be upregulated in hypertrophic tonsil tissue, suggesting that it may play a role in the pathogenesis of inflamed disease tissue.

Type: Primary
Antigen: BPIL3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-100/453
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR