Anti-BPIFB6 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-8425R-CY7
BOSSBS-8425R-CY7EA 900 EUR
BOSSBS-8425R-CY7
Anti-BPIFB6 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Members of the bactericidal/permeability-increasing protein family have antimicrobial properties and bind lipophilic substances, therefore targeting gram-negative bacteria. The bactericidal permeability increasing protein (BPI) is an antibacterial and endotoxin-neutralizing molecule that is abundant in the granules of polymorphonuclear leukocytes (neutrophil granules). Sharing structural and sequence homologies with BPI, BPIL3 (Bactericidal/permeability-increasing protein-like 3) is a 453 amino acid secreted protein that contains the family’s common conserved feature of two cysteine residues that are critical for protein function. While BPIL3 is primarily expressed at low levels in tonsils, it has been found to be upregulated in hypertrophic tonsil tissue, suggesting that it may play a role in the pathogenesis of inflamed disease tissue.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BPIFB6
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR