Anti-ATF2 Thr55 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-8450R-A680
BOSSBS-8450R-A680EA 893 EUR
BOSSBS-8450R-A680
Anti-ATF2 Thr55 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
This gene encodes a transcription factor that is a member of the leucine zipper family of DNA binding proteins. This protein binds to the cAMP-responsive element (CRE), an octameric palindrome. It forms a homodimer or a heterodimer with c-Jun and stimulates CRE-dependent transcription. This protein is also a histone acetyltransferase (HAT) that specifically acetylates histones H2B and H4 in vitro; thus it may represent a class of sequence-specific factors that activate transcription by direct effects on chromatin components. Several alternatively spliced transcript variants have been found for this gene.
Type: Primary
Antigen: ATF2 Thr55
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen