Anti-FNDC5 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-8486R
BOSSBS-8486REA 718 EUR
BOSSBS-8486R
Anti-FNDC5 Rabbit Polyclonal Antibody
Antikörper
Irisin: Contrary to mouse, may not be involved in the beneficial effects of muscular exercise, nor in the induction of browning of human white adipose tissue.

WB(1:100-1000), FCM(1:20-100), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: FNDC5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR