Anti-FNDC5 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-8486R-CY5.5
BOSSBS-8486R-CY5.5EA 897 EUR
BOSSBS-8486R-CY5.5
Anti-FNDC5 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Irisin: Contrary to mouse, may not be involved in the beneficial effects of muscular exercise, nor in the induction of browning of human white adipose tissue.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: FNDC5
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR