Anti-XRCC4 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-8510R-CY7
BOSSBS-8510R-CY7EA 893 EUR
BOSSBS-8510R-CY7
Anti-XRCC4 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Involved in DNA non-homologous end joining (NHEJ) required for double-strand break repair and V(D)J recombination. Binds to DNA and to DNA ligase IV (LIG4). The LIG4-XRCC4 complex is responsible for the NHEJ ligation step, and XRCC4 enhances the joining activity of LIG4. Binding of the LIG4-XRCC4 complex to DNA ends is dependent on the assembly of the DNA-dependent protein kinase complex DNA-PK to these DNA ends.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: XRCC4
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR