Anti-SDHC Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-8530R-CY5
BOSSBS-8530R-CY5EA 900 EUR
BOSSBS-8530R-CY5
Anti-SDHC Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Membrane-anchoring subunit of succinate dehydrogenase (SDH) that is involved in complex II of the mitochondrial electron transport chain and is responsible for transferring electrons from succinate to ubiquinone (coenzyme Q).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SDHC
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR