Anti-SDHC Rabbit Polyclonal Antibody (Cy5.5®)
Lieferant: Bioss
BS-8530R-CY5.5
BOSSBS-8530R-CY5.5EA 900 EUR
BOSSBS-8530R-CY5.5
Anti-SDHC Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Membrane-anchoring subunit of succinate dehydrogenase (SDH) that is involved in complex II of the mitochondrial electron transport chain and is responsible for transferring electrons from succinate to ubiquinone (coenzyme Q).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SDHC
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human