Anti-Prostaglandin E Receptor EP4 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-8538R
BOSSBS-8538REA 757 EUR
BOSSBS-8538R
Anti-Prostaglandin E Receptor EP4 Rabbit Polyclonal Antibody
Antikörper
Receptor for prostaglandin E2 (PGE2). The activity of this receptor is mediated by G(s) proteins that stimulate adenylate cyclase. Has a relaxing effect on smooth muscle. May play an important role in regulating renal hemodynamics, intestinal epithelial transport, adrenal aldosterone secretion, and uterine function.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: Prostaglandin E Receptor EP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR