Anti-TLR5 Rabbit Polyclonal Antibody (Cy7®)
Lieferant: Bioss
BS-8539R-CY7
BOSSBS-8539R-CY7EA 900 EUR
BOSSBS-8539R-CY7
Anti-TLR5 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Luciferase from the firefly has become one of the more widely used reporter proteins for the study of gene expression. Luciferase catalyzes a bioluminescent reaction which requires the substrate luciferin as well as Mg2+ and ATP. Mixing these reagents with the cell extract containing luciferase, results in a flash of light that decays rapidly. This light can be detected by a luminometer. The total light emission is proportional to the luciferase activity of the sample.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TLR5
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other Species