Anti-OLIG1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Lieferant: Bioss
BS-8548R-A488
BOSSBS-8548R-A488EA 893 EUR
BOSSBS-8548R-A488
Anti-OLIG1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
The oligodendrocyte lineage-specific basic helix-loop-helix (OLIG) family of transcription factors include OLIG1-OLIG3, which differ in tissue expression. OLIG1 and OLIG2 are specifically expressed in nervous tissue as gene regulators of oligodendrogenesis. OLIG2 is more widely expressed in embryonic brain than OLIG1, while OLIG3 is primarily expressed in non-neural tissues. OLIG1 and OLIG2 interact with the Nkx-2.2 homeodomain protein, which is responsible for directing ventral neuronal patterning in response to graded Sonic hedgehog signaling in the embryonic neural tube. TheseMore Product Information