Anti-NMDAR2B Tyr1474 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-8566R-A680
BOSSBS-8566R-A680EA 922 EUR
BOSSBS-8566R-A680
Anti-NMDAR2B Tyr1474 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
N-methyl-D-aspartate (NMDA) receptors are a class of ionotropic glutamate receptors. NMDA receptor channel has been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDA receptor channels are heteromers composed of three different subunits: NR1 (GRIN1), NR2 (GRIN2A, GRIN2B, GRIN2C, or GRIN2D) and NR3 (GRIN3A or GRIN3B). The NR2 subunit acts as the agonist binding site for glutamate. This receptor is the predominant excitatory neurotransmitter receptor in the mammalian brain.

Type: Primary
Antigen: NMDAR2B Tyr1474
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR