Anti-DHRS2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-8573R-A680
BOSSBS-8573R-A680EA 1350 EUR
BOSSBS-8573R-A680
Anti-DHRS2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Displays NADPH-dependent dicarbonyl reductase activity in vitro with 3,4-Hexanedione, 2,3-Heptanedione and 1-Phenyl-1,2-propanedione as substrates. No reductase activity is displayed in vitro with steroids, retinoids and sugars as substrates. Attenuates MDM2-mediated p53/TP53 degradation, leading to p53/TP53 stabilisation and increased transcription activity, resulting in the accumulation of MDM2 and CDKN1A/p21.

Type: Primary
Antigen: DHRS2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 21-120/258
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR