Anti-GRAMD3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-8580R-A555
BOSSBS-8580R-A555EA 1350 EUR
BOSSBS-8580R-A555
Anti-GRAMD3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
GRAMD3 is a 432 amino acid protein that contains one GRAM domain and is encoded by a gene that maps to human chromosome 5q23.2. With 181 million base pairs encoding around 1,000 genes, chromosome 5 is about 6% of human genomic DNA. It is associated with Cockayne syndrome through the ERCC8 gene and familial adenomatous polyposis through the adenomatous polyposis coli (APC) tumor suppressor gene. Treacher Collins syndrome is also chromosome 5 associated and is caused by insertions or deletions within the TCOF1 gene. Deletion of the p arm of chromosome 5 leads to Cri du chat syndrome. Deletion of 5q or chromosome 5 altogether is common in therapy-related acute myelogenous leukemias and myelodysplastic syndrome.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GRAMD3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR