Anti-USP22 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-8611R-CY7
BOSSBS-8611R-CY7EA 900 EUR
BOSSBS-8611R-CY7
Anti-USP22 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Histone deubiquitinating component of the transcription regulatory histone acetylation (HAT) complex SAGA. Catalyzes the deubiquitination of both histones H2A and H2B, thereby acting as a coactivator. Recruited to specific gene promoters by activators such as MYC, where it is required for transcription. Required for nuclear receptor-mediated transactivation and cell cycle progression.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: USP22
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR