Anti-C2 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-8613R-CY3
BOSSBS-8613R-CY3EA 900 EUR
BOSSBS-8613R-CY3
Anti-C2 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Component C2 which is part of the classical pathway of the complement system is cleaved by activated factor C1 into two fragments: C2b and C2a. C2a, a serine protease, then combines with complement factor C4b to generate the C3 or C5 convertase.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: C2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR