Anti-RNASE3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-8615R-A488
BOSSBS-8615R-A488EA 890 EUR
BOSSBS-8615R-A488
Anti-RNASE3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Cytotoxin and helminthotoxin with low-efficiency ribonuclease activity. Possesses a wide variety of biological activities. Exhibits antibacterial activity, including cytoplasmic membrane depolarization of preferentially Gram-negative, but also Gram-positive strains. Promotes E.coli outer membrane detachment, alteration of the overall cell shape and partial loss of cell content.

Recommended Dilutions: IF(IHC-P): 1:50-200; IF(IHC-F)(1:50-200

Type: Primary
Antigen: RNASE3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR