Anti-ECP Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-8615R-A680
BOSSBS-8615R-A680EA 890 EUR
BOSSBS-8615R-A680
Anti-ECP Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Cytotoxin and helminthotoxin with low-efficiency ribonuclease activity. Possesses a wide variety of biological activities. Exhibits antibacterial activity, including cytoplasmic membrane depolarisation of preferentially Gram-negative, but also Gram-positive strains. Promotes E.coli outer membrane detachment, alteration of the overall cell shape and partial loss of cell content.

Type: Primary
Antigen: Eosinophil cationic protein
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 45-160/160
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR