Anti-S. enteritidis H Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-8670R-CY5.5
BOSSBS-8670R-CY5.5EA 900 EUR
BOSSBS-8670R-CY5.5
Anti-S. enteritidis H Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
The genus Salmonella is a member of the family Enterobacteriaceae. The genus is composed of Gram-negative bacilli that are facultative and flagellated (motile). Salmonellae possess 3 major antigens; the "H" or flagellar antigen (phase 1 & 2), the "O" or somatic antigen (part of the LPS moiety) and the "Vi" or capsular antigen (referred to as "K" in other Enterobacteriaceae). Salmonellae also possess the LPS endotoxin characteristic of Gram-negative bacteria. This LPS is composed of an "O" polysaccharide ("O" antigen) an "R" core and the endotoxic inner "Lipid A". Endotoxins evoke fever and can activate complement, kinin and clotting factors. The commonest Salmonella serotype associated with food borne infections in humans is Salmonella enteriditis and in particular phage type 4 (PT4). Salmonella Enteriditis bacteria may be found in the intestinal tracts of livestock, poultry, dogs, cats and other warm-blooded animals. This strain is only one of about 2,000 kinds of Salmonella bacteria; it is often associated with poultry and eggs.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Salmonella enteritidis H
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR