Anti-SMS Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-8681R
BOSSBS-8681REA 725 EUR
BOSSBS-8681R
Anti-SMS Rabbit Polyclonal Antibody
Antikörper
Spermine synthase catalyzes the production of spermine from spermidine. Spermine, a polyamine ubiquitously present in most organisms, is essential for normal cell growth and differentiation. Because absence of spermine increases sensitivity of cells to anti-tumor agents, spermine synthase (and other polyamine biosynthesis) is an attractive target for anti-neoplastic therapy.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: SMS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR