Anti-SMS Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-8681R-HRP
BOSSBS-8681R-HRPEA 900 EUR
BOSSBS-8681R-HRP
Anti-SMS Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Spermine synthase catalyzes the production of spermine from spermidine. Spermine, a polyamine ubiquitously present in most organisms, is essential for normal cell growth and differentiation. Because absence of spermine increases sensitivity of cells to anti-tumor agents, spermine synthase (and other polyamine biosynthesis) is an attractive target for anti-neoplastic therapy.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: SMS
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR