Anti-CDK4 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-8682R-CY7
BOSSBS-8682R-CY7EA 900 EUR
BOSSBS-8682R-CY7
Anti-CDK4 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
The activation of RaP1 by cAMP is independent of PKA and is mediated by recently discovered family of guanine nucleotide exchange factors (GEFs) called cAMP-GEFs or Epacs. The Epac signaling therefore represents a novel mechanism for cAMP signaling with in the cAMP cascade. There are 2 members of the Epac family, Epac1 and Epac 2. Both proteins are multidomain proteins containing an autoinhibitory cAMP-binding domain that inhibits the catalytic region and a DEP domain (dishevelled, Egl-10 and pleckstrin homology domain) targeting the membrane anchors. EPAC2 has an additional cAMP-binding site in its N-terminus that binds cAMP with low affinity. EPAC1 mRNA is broadly expressed, with particularly high levels occurring in the thyroid, ovary, kidney and certain brain regions, whereas expression of EPAC2 mRNA appears to be restricted to the brain and adrenal glands. Epac 1 and Epac 2 also interact with light chain 2 (LC2) or MAP1A that serves as a scaffolding structure to stabilize the signal transduction complex. The Epac 1-selective were generated against unique antigenic sequences form near N-terminus and between RasGEFN and Ras GEF domains. The to Epac 1are affinity purified over immobilized antigen based chromatography.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CDK4
Clonality: Polyclonal
Clone: 0
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR