Anti-Viperin Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-8718R-A350
BOSSBS-8718R-A350EA 893 EUR
BOSSBS-8718R-A350
Anti-Viperin Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
RSAD2 is a 361 amino acid protein that is involved in antiviral defense against pathogens such as Hep C, Cytomegalovirus and HIV-1. Localized to the cytosolic side of the endoplasmic reticulum and relocated to the Golgi apparatus upon viral infection, RSAD2 is thought to prevent viral budding by disrupting lipid rafts at the plasma membrane and supporting the Interferon-induced antiviral state of the cell. Additionally, RSAD2 can bind to and inactivate FDPS (an enzyme that is crucial for the synthesis of cholesterol and geranylated and farnesylated proteins), thereby playing a role in lipid synthesis. Overexpression of RSAD2 leads to abnormal lipid accumulation that is associated with atherosclerosis, a chronic inflammatory disease characterized by hardened arteries.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Viperin
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR